Assay Details
| Gene Symbol: 1700093C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007047.1 | - | 1 | 455 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 963 | Zfp963 | Anti-sense | Chr. 8 - 69741639 - 69749962 on NCBI Build GRCm38 | ||
| Downstream | zinc finger protein 866 | Zfp866 |
9830167H18Rik, AI875089, C85843, D330038O06Rik |
10.7kb | Anti-sense | Chr. 8 - 69761324 - 69774911 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700093C20Rik
Genomic Map