Assay Details
| Gene Symbol: 1700101I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045270.1 | 1 - 2 | 47 | 61 | ||
| GenBank mRNA | AK007107.1 | - | 1 - 2 | 36 | 61 | |
| AK015489.1 | - | 1 - 2 | 48 | 61 | ||
| BC099571.1 | - | 1 - 2 | 47 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | gamma-aminobutyric acid (GABA) A receptor-associated protein-like 1 | Gabarapl1 |
3110025G09Rik, 9130422N19Rik, AI196471, Apg8l, Atg8l, GECI, MNCb-0091 |
Anti-sense | Chr. 6 - 129533192 - 129542331 on NCBI Build GRCm38 | |
| Downstream | killer cell lectin-like receptor family E member 1 | Klre1 |
Klre-1, NKG2I |
25.8kb | Anti-sense | Chr. 6 - 129559032 - 129585827 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700101I11Rik
Genomic Map