Assay Details
| Gene Symbol: 1700102F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007111.2 | - | 1 | 451 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
capping protein (actin filament) muscle Z-line, alpha 3 |
Capza3 |
510-4, Cappa3, Gsg3, Tex8, repro32 |
-266.3kb | Sense | Chr. 6 - 140041525 - 140042789 on NCBI Build GRCm38 |
| Downstream |
pleckstrin homology domain containing, family A member 5 |
Plekha5 |
2810431N21Rik, AI428202, AK129423, AMH-Cre, Ayu21-9, Gt(pU21)9Imeg, Pepp2, Tg(AMH-cre)1Flor |
113.9kb | Sense | Chr. 6 - 140423639 - 140597110 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700102F20Rik
Genomic Map