Assay Details
| Gene Symbol: 1700008B11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007127.1 | - | 1 | 49 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 7031 | Mir7031 |
Gm27859, mmu-mir-7031 |
-15.9kb | Anti-sense | Chr. 5 - 121819209 - 121819273 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 109, member A |
Fam109a |
A230106M15Rik, AU017694, IPIP27A, Ses1 |
13.3kb | Sense | Chr. 5 - 121849000 - 121854606 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700008B11Rik
Genomic Map