Assay Details
| Gene Symbol: 1700108I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007136.1 | - | 1 | 157 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | protein tyrosine phosphatase 4a2 | Ptp4a2 | Prl-2 | -2.9kb | Anti-sense | Chr. 4 - 129820479 - 129850003 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA E330017L17 gene | E330017L17Rik | 53.0kb | Anti-sense | Chr. 4 - 129906226 - 129908996 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700108I11Rik
Genomic Map