Assay Details
| Gene Symbol: 1700112L15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007183.1 | - | 1 | 348 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 113 | Ccdc113 |
4933409I22, BC060957 |
Sense | Chr. 8 - 95534060 - 95558888 on NCBI Build GRCm38 | |
| Downstream |
protease, serine 54 |
Prss54 |
4931432M23Rik, Klkbl4 |
6.8kb | Anti-sense | Chr. 8 - 95559067 - 95576330 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700112L15Rik
Genomic Map