Assay Details
| Gene Symbol: 1700124L16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007271.1 | - | 1 | 350 | 73 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BP759901.1 | - | 1 | Chr. 6 - 83761756 - 83762288 [+] on Build GRCm38 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
ATPase, H+ transporting, lysosomal V1 subunit B1 |
Atp6v1b1 |
AW208839, Atp6b1, D630003L15, D630030L16Rik, D630039P21Rik, Vpp-3, Vpp3 |
-2.9kb | Anti-sense | Chr. 6 - 83743017 - 83758810 on NCBI Build GRCm38 |
| Downstream | ankyrin repeat domain 53 | Ankrd53 | 4930564N15Rik | 153.0 | Anti-sense | Chr. 6 - 83762646 - 83768326 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1700124L16Rik
Genomic Map