Assay Details
| Gene Symbol: 1810044K17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007778.1 | - | 1 - 2 | 294 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1810053B23 gene | 1810053B23Rik |
1810008L18Rik, 1810056K14Rik |
-8.7kb | Anti-sense | Chr. 16 - 93343716 - 93359543 on NCBI Build GRCm38 |
| Downstream | microRNA 802 | Mir802 |
Mirn802, mir-802, mmu-mir-802 |
691.0 | Sense | Chr. 16 - 93369720 - 93369816 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

1810044K17Rik
Genomic Map