Assay Details
| Gene Symbol: 2310075K07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007865.1 | - | 1 | 445 | 104 | |
| AK010182.1 | - | 1 | 550 | 104 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930431F10 gene | 4930431F10Rik | -327.5kb | Sense | Chr. 10 - 44730777 - 44738900 on NCBI Build GRCm38 | |
| Downstream | prolyl endopeptidase | Prep |
AI047692, AI450383, D10Wsu136e, PEP, Pop |
240.0 | Anti-sense | Chr. 10 - 45067214 - 45159981 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310075K07Rik
Genomic Map