Assay Details
| Gene Symbol: 1810058N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007900.1 | - | 1 | 545 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | OVO homolog-like 1 (Drosophila) | Ovol1 |
BB147136, Ovo1, movo1 |
-477.0 | Anti-sense | Chr. 19 - 5549137 - 5560575 on NCBI Build GRCm38 |
| Downstream |
ribonuclease H2, subunit C |
Rnaseh2c |
1500026D16Rik, AYP1 |
40.2kb | Sense | Chr. 19 - 5601873 - 5602959 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1810058N15Rik
Genomic Map