Assay Details
| Gene Symbol: 1810073G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK007969.1 | - | 1 | 1549 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pancreatic lipase related protein 1 | Pnliprp1 | Plrp1 | Sense | Chr. 19 - 58728887 - 58744169 on NCBI Build GRCm38 | |
| Downstream | pancreatic lipase-related protein 2 | Pnliprp2 | PLRP2 | 16.0kb | Sense | Chr. 19 - 58759723 - 58777534 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

1810073G21Rik
Genomic Map