Assay Details
| Gene Symbol: 2010015M23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008259.1 | - | 1 | 152 | 96 | |
| Gene Symbol: Srsf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078167.2 | NP_001071635.1 | 1 | 96 | ||
| NM_173374.4 | NP_775550.2 | 1 | 96 | |||
| GenBank mRNA | CF739274.1 | - | 1 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | dynein light chain LC8-type 2 | Dynll2 |
1700064A15Rik, 6720463E02Rik, C87222, DLC8, DLC8b, Dlc2 |
-48.5kb | Sense | Chr. 11 - 87979525 - 87998298 on NCBI Build GRCm38 |
| Downstream | vascular endothelial zinc finger 1 | Vezf1 |
AI848691, AW046909, db1 |
20.7kb | Anti-sense | Chr. 11 - 88068256 - 88084729 on NCBI Build GRCm38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001078167.2, NM_173374.4 transcripts.
Set Membership: |
|

2010015M23Rik
Genomic Map