Assay Details
| Gene Symbol: 2010205J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008456.1 | - | 1 | 151 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphorylase kinase beta | Phkb | AI463271 | Sense | Chr. 8 - 85840984 - 86064254 on NCBI Build GRCm38 | |
| Downstream |
ATP-binding cassette, sub-family C (CFTR/MRP), member 12 |
Abcc12 |
4930467B22Rik, MRP9 |
440.3kb | Anti-sense | Chr. 8 - 86504576 - 86580690 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2010205J10Rik
Genomic Map