Assay Details
| Gene Symbol: 2210402A03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008796.1 | - | 1 | 1343 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
prenyl (solanesyl) diphosphate synthase, subunit 1 |
Pdss1 |
2610203G20Rik, 2700031G06Rik, TPT, TPT 1, Tprt, mDLP1, mSPS1 |
-51.2kb | Anti-sense | Chr. 2 - 22893560 - 22940266 on NCBI Build GRCm38 |
| Downstream | acyl-Coenzyme A binding domain containing 5 | Acbd5 | 1300014E15Rik | 74.2kb | Anti-sense | Chr. 2 - 23067700 - 23114513 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2210402A03Rik
Genomic Map