Assay Details
| Gene Symbol: 2210414I22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK008926.1 | - | 1 | 347 | 80 | |
| AK132110.1 | - | 1 | 476 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
zinc finger protein 335, opposite strand |
Zfp335os |
Gm11458, OTTMUSG00000001112 |
Sense | Chr. 2 - 164911349 - 164921265 on NCBI Build GRCm38 | |
| Downstream | matrix metallopeptidase 9 | Mmp9 |
AW743869, B/MMP9, Clg4b, MMP-9, pro-MMP-9 |
28.0kb | Sense | Chr. 2 - 164940744 - 164955850 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2210414I22Rik
Genomic Map