Assay Details
| Gene Symbol: 2310007H11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009208.1 | - | 1 | 453 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
phosphatidylinositol glycan anchor biosynthesis, class T |
Pigt |
2510012P17Rik, 4930534E15Rik, CGI-06, NDAP, Ndap7 |
-13.9kb | Sense | Chr. 2 - 164497511 - 164508301 on NCBI Build GRCm38 |
| Downstream | predicted gene 14317 | Gm14317 | OTTMUSG00000016294 | 25.7kb | Sense | Chr. 2 - 164549485 - 164559748 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310007H11Rik
Genomic Map