Assay Details
| Gene Symbol: 2310015D24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037997.1 | 4 | 2060 | 79 | ||
| GenBank mRNA | AK009355.1 | - | 1 | 746 | 79 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6365 | Mir6365 |
Gm27683, mmu-mir-6365 |
-43.0kb | Sense | Chr. 16 - 13471080 - 13471180 on NCBI Build GRCm38 |
| Downstream | poly(A)-specific ribonuclease (deadenylation nuclease) | Parn |
1200003I18Rik, DAN |
22.2kb | Sense | Chr. 16 - 13537960 - 13668196 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310015D24Rik
Genomic Map