Assay Details
| Gene Symbol: Pcif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009480.1 | - | 1 | 351 | 133 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phospholipid transfer protein | Pltp |
Bpife, OD107 |
-25.1kb | Anti-sense | Chr. 2 - 164839518 - 164857708 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 335 | Zfp335 |
1810045J01Rik, NIF-1, Nif1, Znf335 |
8.7kb | Anti-sense | Chr. 2 - 164891882 - 164911750 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcif1
Genomic Map