Assay Details
| Gene Symbol: Pcgf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009638.1 | - | 48 | 85 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | major facilitator superfamily domain containing 7A | Mfsd7a |
4732482E20Rik, Mfsd7 |
-52.8kb | Anti-sense | Chr. 5 - 108441052 - 108450260 on NCBI Build GRCm38 |
| Downstream | complexin 1 | Cplx1 | 921-S | 14.8kb | Anti-sense | Chr. 5 - 108518554 - 108550027 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pcgf3
Genomic Map