Assay Details
| Gene Symbol: 2310045N14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK009831.1 | - | 1 | 753 | 115 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
myosin light chain kinase family, member 4 |
Mylk4 | EG238564 | Sense | Chr. 13 - 32700827 - 32806771 on NCBI Build GRCm38 | |
| Downstream | Werner helicase interacting protein 1 | Wrnip1 |
4833444L21Rik, WHIP, Wrnip |
75.1kb | Anti-sense | Chr. 13 - 32802030 - 32822610 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2310045N14Rik
Genomic Map