Assay Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020870.1 | - | 1 | 251 | 84 | |
| AK052686.1 | - | 6 | 2009 | 84 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 15713 | Gm15713 | OTTMUSG00000025617 | -158.2kb | Anti-sense | Chr. 16 - 43410518 - 43420196 on NCBI Build GRCm38 |
| Downstream | microRNA 568 | Mir568 |
Mirn568, mmu-mir-568 |
61.8kb | Sense | Chr. 16 - 43640655 - 43640737 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Zbtb20
Genomic Map