Assay Details
| Gene Symbol: A930026C06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020892.1 | - | 1 | 348 | 83 | |
| AK048940.1 | - | 1 | 350 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptotagmin IX | Syt9 | Sense | Chr. 7 - 107370704 - 107548656 on NCBI Build GRCm38 | ||
| Downstream | olfactomedin-like 1 | Olfml1 |
6720478C22, AV321997, BC047207, MVAL564, ONT2, mONT2 |
49.3kb | Sense | Chr. 7 - 107567433 - 107591365 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930026C06Rik
Genomic Map