Assay Details
| Gene Symbol: A930041H05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020950.1 | - | 1 | 643 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
poly (ADP-ribose) polymerase family, member 8 |
Parp8 |
2810430O08Rik, ARTD16, D13Ertd275e |
Sense | Chr. 13 - 116850842 - 117025913 on NCBI Build GRCm38 | |
| Downstream | predicted gene 6416 | Gm6416 | 277.8kb | Anti-sense | Chr. 13 - 117130025 - 117135884 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A930041H05Rik
Genomic Map