Assay Details
| Gene Symbol: Rapgef4it | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK020960.1 | - | 1 | 846 | 61 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Rap guanine nucleotide exchange factor (GEF) 4 | Rapgef4 |
1300003D15Rik, 5730402K07Rik, 6330581N18Rik, EPAC 2, Epac-2, Epac2, mKIAA4040 |
Sense | Chr. 2 - 71981215 - 72257474 on NCBI Build GRCm38 | |
| Downstream | uncharacterized LOC105244195 | LOC105244195 | 138.0kb | Anti-sense | Chr. 2 - 72153177 - 72234068 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Rapgef4it
Genomic Map