Assay Details
| Gene Symbol: C030001C17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021036.1 | - | 1 | 645 | 83 | |
| CK781252.1 | - | 1 | 451 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cysteine-rich perinuclear theca 14 | Cypt14 | -1.8mb | Anti-sense | Chr. X - 39862919 - 39863604 on NCBI Build GRCm38 | |
| Downstream | THO complex 2 | Thoc2 |
6330441O12Rik, D130005M13Rik, Gm1139, Gm1793, Tho2 |
166.5kb | Anti-sense | Chr. X - 41794992 - 41911924 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030001C17Rik
Genomic Map