Assay Details
| Gene Symbol: C030011I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021065.1 | - | 1 | 445 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | predicted gene 4719 | Gm4719 | -1.4mb | Anti-sense | Chr. 17 - 89379241 - 89384993 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930480K15 gene | 4930480K15Rik | 95.7kb | Anti-sense | Chr. 17 - 90861169 - 90867102 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030011I16Rik
Genomic Map