Assay Details
| Gene Symbol: C030014C12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021079.1 | - | 1 | 751 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | myelin transcription factor 1-like | Myt1l |
2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106 |
Sense | Chr. 12 - 29528311 - 29923216 on NCBI Build GRCm38 | |
| Downstream | peroxidasin homolog (Drosophila) | Pxdn |
2310075M15Rik, C85409, E330004E07, VPO1, mKIAA0230 |
136.7kb | Sense | Chr. 12 - 29937877 - 30017658 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030014C12Rik
Genomic Map