Assay Details
| Gene Symbol: C030032G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021114.1 | - | 1 | 351 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
nardilysin, N-arginine dibasic convertase, NRD convertase 1 |
Nrd1 |
2600011I06Rik, AI875733, NRD-C |
-41.2kb | Anti-sense | Chr. 4 - 109000615 - 109061771 on NCBI Build GRCm38 |
| Downstream | calreticulin 4 | Calr4 |
4933403L16Rik, AU017583 |
101.3kb | Anti-sense | Chr. 4 - 109204779 - 109255138 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C030032G21Rik
Genomic Map