Assay Details
| Gene Symbol: Mbd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021136.1 | - | 549 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Downstream | enhancer of polycomb homolog 2 (Drosophila) | Epc2 |
5830499L14, D2Ertd694e |
131.8kb | Anti-sense | Chr. 2 - 49451486 - 49551942 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mbd5
Genomic Map