Assay Details
| Gene Symbol: Prkce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011104.3 | NP_035234.1 | 15 | 4951 | 79 | |
| GenBank mRNA | AK017901.1 | - | 1 | 582 | 79 | |
| AK021153.1 | - | 1 | 152 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 2010106C02 gene | 2010106C02Rik | -369.3kb | Anti-sense | Chr. 17 - 86285167 - 86287178 on NCBI Build GRCm38 | |
| Downstream | endothelial PAS domain protein 1 | Epas1 |
HIF-2alpha, HIF2A, HLF, HRF, MOP2, bHLHe73 |
97.0kb | Sense | Chr. 17 - 86753864 - 86833410 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Prkce
Genomic Map