Assay Details
| Gene Symbol: Pde8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021156.1 | - | 1 | 546 | 92 | |
| AK034510.1 | - | 1 | 92 | |||
| AK052428.1 | - | 1 | 92 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | WD repeat domain 41 | Wdr41 |
B830029I03Rik, MSTP048 |
-6.3kb | Anti-sense | Chr. 13 - 94976213 - 95023316 on NCBI Build GRCm38 |
| Downstream |
zinc finger, BED type containing 3 |
Zbed3 |
0610037K01Rik, 2610005H11Rik, AU018975, AU024588 |
293.3kb | Anti-sense | Chr. 13 - 95323605 - 95337842 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pde8b
Genomic Map