Assay Details
| Gene Symbol: Jakmip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021217.1 | - | 1 | 644 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Wolfram syndrome 1 homolog (human) | Wfs1 |
AI481085, wolframin |
-102.1kb | Anti-sense | Chr. 5 - 36966104 - 36988982 on NCBI Build GRCm38 |
| Downstream | predicted gene 1043 | Gm1043 | 61.1kb | Sense | Chr. 5 - 37153129 - 37234484 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Jakmip1
Genomic Map