Assay Details
| Gene Symbol: C430050I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021264.1 | - | 1 | 749 | 103 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | DV062400.1 | - | 238 | Chr. - - [-] on Build | 103 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor of activated T cells 5 | Nfat5 |
AI225870, B130038B15Rik, CAG-8, CAG80, NFATL1, OREBP, TonEBP, mKIAA0827, nfatz |
Sense | Chr. 8 - 107293000 - 107379517 on NCBI Build GRCm38 | |
| Downstream |
NAD(P)H dehydrogenase, quinone 1 |
Nqo1 |
AV001255, Dia4, Dtd, Nmo-1, Nmo1, Nmor1, Ox-1, Ox1, Qr1 |
51.8kb | Anti-sense | Chr. 8 - 107388225 - 107403205 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

C430050I21Rik
Genomic Map