Assay Details
| Gene Symbol: B430319G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029474.1 | 1 | 999 | 70 | ||
| GenBank mRNA | AK021297.1 | - | 1 | 243 | 70 | |
| AK044362.1 | - | 1 | 482 | 70 | ||
| AK046705.1 | - | 1 | 1000 | 70 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phospholipid scramblase 4 | Plscr4 | AV245873 | -48.7kb | Anti-sense | Chr. 9 - 92457372 - 92492516 on NCBI Build GRCm38 |
| Downstream |
procollagen lysine, 2-oxoglutarate 5-dioxygenase 2 |
Plod2 |
D530025C14Rik, LH2, Plod-2 |
111.0 | Anti-sense | Chr. 9 - 92542223 - 92608428 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

B430319G15Rik
Genomic Map