Assay Details
| Gene Symbol: D730050C22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK021359.1 | - | 1 | 946 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thyroid hormone receptor associated protein 3 | Thrap3 |
9330151F09Rik, B230333E16Rik, Trap150 |
-168.0 | Anti-sense | Chr. 4 - 126164082 - 126202710 on NCBI Build GRCm38 |
| Downstream | MAP7 domain containing 1 | Map7d1 |
AV028413, BC019977, Mtap7d1, Parcc1, Rprc1, mKIAA1187 |
28.2kb | Anti-sense | Chr. 4 - 126232167 - 126256417 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

D730050C22Rik
Genomic Map