Assay Details
| Gene Symbol: Luc7l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK042182.1 | - | 1 | 503 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 1110001J03 gene | 1110001J03Rik | Fmc1 | -58.6kb | Sense | Chr. 6 - 38534861 - 38539449 on NCBI Build GRCm38 |
| Downstream |
killer cell lectin-like receptor subfamily G, member 2 |
Klrg2 | 2310020F24Rik | 22.7kb | Anti-sense | Chr. 6 - 38621414 - 38637706 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Luc7l2
Genomic Map