Assay Details
| Gene Symbol: 4122401K19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014465.1 | - | 1 | 2350 | 75 | |
| AK029213.1 | - | 1 | 1051 | 75 | ||
| AV243710.2 | - | 1 | 416 | 75 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA A330040F15 gene | A330040F15Rik | -486.0 | Anti-sense | Chr. 19 - 12585868 - 12596566 on NCBI Build GRCm38 | |
| Downstream | predicted gene 4952 | Gm4952 | EG240549 | 1.7kb | Sense | Chr. 19 - 12599984 - 12627616 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4122401K19Rik
Genomic Map