Assay Details
| Gene Symbol: E130014H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029413.1 | - | 1 | 350 | 80 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 31282 |
Gm31282 | -162.6kb | Anti-sense | Chr. 15 - 8468493 - 8502882 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31340 |
Gm31340 | 43.8kb | Anti-sense | Chr. 15 - 8711026 - 8864483 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E130014H10Rik
Genomic Map