Assay Details
Gene Symbol: 4933400F21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015540.1 | 2 - 3 | 322 | 75 | ||
GenBank mRNA | AK016592.1 | - | 2 - 3 | 324 | 75 | |
AK029704.1 | - | 2 - 3 | 327 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA C030007H22 gene | C030007H22Rik | -273.8kb | Sense | Chr. 1 - 89360342 - 89406133 on NCBI Build GRCm38 | |
Downstream | gastrulation brain homeobox 2 | Gbx2 |
D130058E05Rik, Gbx-2, MMoxA, Stra7 |
246.7kb | Sense | Chr. 1 - 89927962 - 89931176 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
