Assay Details
| Gene Symbol: Chmp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030270.1 | - | 1 | 2349 | 72 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | R3H domain and coiled-coil containing 1 | R3hcc1 |
1700020M16Rik, AA407796 |
-13.6kb | Anti-sense | Chr. 14 - 69697304 - 69714809 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930480K23 gene | 4930480K23Rik | 1.9kb | Sense | Chr. 14 - 69732784 - 69767113 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Chmp7
Genomic Map