Assay Details
| Gene Symbol: 6430537K16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030418.1 | - | 1 | 1947 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ephrin A5 | Efna5 |
AL-1, AV158822, EFL-5, Ephrin-A5, Epl7, LERK-7, RAGS |
Sense | Chr. 17 - 62602957 - 62881317 on NCBI Build GRCm38 | |
| Downstream | F-box and leucine-rich repeat protein 17 | Fbxl17 |
6330576B01Rik, AI452053, BB073797, C130023C01Rik, Fbx13, Fbxo13 |
196.5kb | Sense | Chr. 17 - 63045949 - 63504042 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6430537K16Rik
Genomic Map