Assay Details
Gene Symbol: Grm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK032068.1 | - | 1 | 1950 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 20387 | Gm20387 | -918.1kb | Anti-sense | Chr. 6 - 110639752 - 110645916 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 1700054K19 gene | 1700054K19Rik | 643.5kb | Sense | Chr. 6 - 112210708 - 112212519 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
