Assay Details
| Gene Symbol: 2900075N08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK032471.1 | - | 1 | 1651 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuroligin 3 | Nlgn3 |
A230085M13Rik, HNL3, NL3 |
-2.2kb | Sense | Chr. X - 101299179 - 101321350 on NCBI Build GRCm38 |
| Downstream |
gap junction protein, beta 1 |
Gjb1 |
Cnx32, Cx32 |
50.4kb | Sense | Chr. X - 101376376 - 101385629 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900075N08Rik
Genomic Map