Assay Details
| Gene Symbol: Traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK033718.1 | - | 1 | 2748 | 92 | |
| AK083275.1 | - | 2086 | 92 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | recombination activating gene 1 | Rag1 | Rag-1 | -49.2kb | Anti-sense | Chr. 2 - 101638252 - 101649532 on NCBI Build GRCm38 |
| Downstream | proline rich 5 like | Prr5l |
2600010E01Rik, 4833411O04Rik, Protor-2, Protor2 |
12.6kb | Anti-sense | Chr. 2 - 101714280 - 101883074 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Traf6
Genomic Map