Assay Details
| Gene Symbol: C030029H02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102277.1 | 3 - 4 | 1480 | 104 | ||
| GenBank mRNA | AK034027.1 | - | 3 - 4 | 1482 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 6330420H09 gene | 6330420H09Rik | -424.5kb | Sense | Chr. 7 - 135853682 - 135855976 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA C230079O03 gene | C230079O03Rik | C230079O03 | 13.7kb | Anti-sense | Chr. 7 - 136332216 - 136349415 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 805382 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54799 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 43532 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 241771 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 798891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 614421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 23779 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526697 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 846119 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229285 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 100988 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 38829 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422881 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 411761 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164554 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128079 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150066 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01647022 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01646963 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01646999 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06384043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01646999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06551425 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06212214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558176 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558249 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558253 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558128 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06551434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558015 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06575991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558280 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558295 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557947 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05911161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06096674 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05911159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558009 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558241 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05942131 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558291 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558294 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06535797 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06439582 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558244 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558239 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558246 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557929 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557927 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06409069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558146 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06575999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558273 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557958 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558148 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558117 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06063224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06212219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558283 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558289 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558228 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06531993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558140 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558206 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06587989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06063219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558290 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558262 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06372737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558276 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558221 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06409073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558215 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558213 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558287 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557911 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06535785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558154 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06138598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558002 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557913 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558214 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558270 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558284 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558142 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06109886 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06015611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06109860 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558219 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06409064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558210 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06409074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06096677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06587184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557964 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558220 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06109864 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06094436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557966 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06090305 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558121 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558028 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06191950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557939 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558247 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558252 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06109883 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557973 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06082136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05896604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558225 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557975 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25558272 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25557950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576000 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0700017264.mm.1 | TC0700001971.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017264.mm.1 | NR_102277.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017266.mm.1 | TC0700001971.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017266.mm.1 | NR_102277.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700017265.mm.1 | TC0700001971.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700017265.mm.1 | NR_102277.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 104120500 | - | - |

C030029H02Rik
Genomic Map