Assay Details
| Gene Symbol: 9430062P05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK034922.1 | - | 1 | 2845 | 102 | |
| BB098221.2 | - | 1 | 191 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mortality factor 4 like 1 | Morf4l1 |
MORFRG15, MRG15, TEG-189, Tex189, mKIAA4002 |
-41.6kb | Sense | Chr. 9 - 90091669 - 90114820 on NCBI Build GRCm38 |
| Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7 |
Adamts7 |
ADAM-TS7, ADAMTS7B |
3.2kb | Anti-sense | Chr. 9 - 90162978 - 90200102 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9430062P05Rik
Genomic Map