Assay Details
Gene Symbol: AW549542 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045702.1 | 3 - 4 | 1322 | 93 | ||
GenBank mRNA | AK035112.1 | - | 1344 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31314 |
Gm31314 | -42.5kb | Sense | Chr. 5 - 119472143 - 119527976 on NCBI Build GRCm38 | |
Downstream |
T-box 3, opposite strand 1 |
Tbx3os1 | Gm2301 | 26.4kb | Sense | Chr. 5 - 119597977 - 119671331 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
