Assay Details
| Gene Symbol: Fam129a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035217.1 | - | 1 | 449 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ring finger protein 2 | Rnf2 |
AI326319, AI450156, AU019207, Ring1B, dinG |
-218.1kb | Anti-sense | Chr. 1 - 151469406 - 151500923 on NCBI Build GRCm38 |
| Downstream |
ER degradation enhancer, mannosidase alpha-like 3 |
Edem3 | 2310050N11Rik | 34.5kb | Sense | Chr. 1 - 151755344 - 151822328 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fam129a
Genomic Map