Assay Details
| Gene Symbol: 9530036O11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015562.1 | 1 | 160 | 61 | ||
| GenBank mRNA | AK035417.1 | - | 1 | 648 | 61 | |
| AK035474.1 | - | 1 | 160 | 61 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sonic hedgehog | Shh |
9530036O11Rik, Dsh, Hhg1, Hx, Hxl3, M100081 |
Anti-sense | Chr. 5 - 28456840 - 28467101 on NCBI Build GRCm38 | |
| Downstream | ring finger protein 32 | Rnf32 |
1700009J01Rik, 2700025B22Rik, 4930542N22Rik, Lmbr2 |
727.9kb | Sense | Chr. 5 - 29195170 - 29225526 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

9530036O11Rik
Genomic Map