Assay Details
| Gene Symbol: 2900022M12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035467.1 | - | 1 | 2648 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chloride intracellular channel 6 | Clic6 |
5730466J16Rik, AL022908, AW045520, CLIC1L |
-143.0kb | Anti-sense | Chr. 16 - 92498147 - 92541241 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 1810053B23 gene | 1810053B23Rik |
1810008L18Rik, 1810056K14Rik |
656.5kb | Sense | Chr. 16 - 93343716 - 93359543 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2900022M12Rik
Genomic Map